Order Entry
Italy
ContactUsLinkComponent
 
 
4-Fluoro-1,3-phenylenediamine ≥98.0% (by GC, titration analysis)
  TCIAF0794-5G
 :  TCI
 :  
 
4-Fluoro-1,3-phenylenediamine ≥98.0% (by GC, titration analysis)
  TCIAF0794-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Brown powder to lump
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

 

 :
 :126.13 g/mol
 :153°C
 :228-426-4
 :GHS07
 :Refrigerator
 :6264-67-1
 :C₆H₇FN₂
 :MFCD00077494