
Formaldehyde Dimethylhydrazone ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2035-89-4
:
Formula:C₃H₈N₂
MDL Number:MFCD01753011
:72.11 g/mol
Boiling Pt:71°C
Melting Pt:–103°C
Storage Temperature:Refrigerator
UN:1993
ADR:3,II
:GHS02
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together