Order Entry
Italy
ContactUsLinkComponent
1,1'-Ferrocenediboronic acid ≥95.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
Irritant
1,1'-Ferrocenediboronic acid ≥95.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
Catalog # TCIAF0664-1G
Supplier:  TCI
CAS Number:  
Irritant
1,1'-Ferrocenediboronic acid ≥95.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
Catalog # TCIAF0664-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (Chelometric Titration):
    min. 95.0 %

Specification Test Results

About this item

 :
 :273.67 g/mol
 :GHS07
CAS Number:32841-83-1
Formula:C₁₀H₁₂B₂FeO₄
MDL Number:MFCD00799542