Order Entry
Italy
ContactUsLinkComponent
2-(2-Fluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane ≥98.0% (by titrimetric analysis)
2-(2-Fluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane ≥98.0% (by titrimetric analysis)
Catalog # TCIAF0531-1G
Supplier:  TCI
CAS Number:  
2-(2-Fluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane ≥98.0% (by titrimetric analysis)
Catalog # TCIAF0531-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :208.04 g/mol
Storage Temperature:Refrigerator
CAS Number:346656-39-1
Formula:C₁₁H₁₄BFO₂
MDL Number:MFCD06200700