2-[2-(Furan-2-yl)vinyl]-4,6-bis(trichloromethyl)-1,3,5-triazine ≥95.0% (by HPLC, total nitrogen)
Catalog # TCIAF0362-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (with Total Nitrogen):min. 95.0 %
- Melting point:140.0 to 144.0 deg-C
Specification Test Results
About this item
Synonyms: 2,4-Bis(trichloromethyl)-6-[2-(furan-2-yl)vinyl]-1,3,5-triazine
:
:407.88 g/mol
CAS Number:154880-05-4
Formula:C₁₁H₅Cl₆N₃O
MDL Number:MFCD00671535