Fumaraldehyde mono(dimethyl acetal) ≥95.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:4093-49-6
:
Formula:C₆H₁₀O₃
MDL Number:MFCD00191439
:130.14 g/mol
:Stabilised with hydroquinone (HQ)
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together