Formyltetrathiafulvalene ≥98.0% (by GC)
Catalog # TCIAF0285-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Dark green to Dark red to Black powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:111.0 to 115.0 deg-C
Specification Test Results
About this item
:
:232.35 g/mol
CAS Number:68128-94-9
Formula:C₇H₄OS₄