
4-Fluoro-N,N-dimethylaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:403-46-3
:
Formula:C₈H₁₀FN
MDL Number:MFCD00059471
Boiling Pt:200°C
:139.17 g/mol
:GHS07
Specification Test Results
- Appearance:White or Colorless to Light orange to Yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:35.0 to 39.0 deg-C
Specification Test Results
Frequently Bought Together