N-Formyl-D-phenylalanine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:59366-89-1
:
Formula:C₁₀H₁₁NO₃
MDL Number:MFCD00063282
:193.20 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:163.0 to 168.0 deg-C
- Specific rotation [a]20/D:-63.0 to -69.0 deg (C=4,EtOH)
- Solubility in EtOH:almost transparency
Specification Test Results
Frequently Bought Together