N-Formylglycine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2491-15-8
:
Formula:C₃H₅NO₃
MDL Number:MFCD00037360
:103.08 g/mol
EINECS:219-649-8
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Water:very faint turbidity
Specification Test Results
Frequently Bought Together