Furil ≥98.0% (by GC)
Catalog # TCIAF0078-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Amber powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:163.0 to 166.0 deg-C
- Solubility in hot EtOH:almost transparency
Specification Test Results
About this item
:
:190.15 g/mol
EINECS:207-766-7
CAS Number:492-94-4
Formula:C₁₀H₆O₄
MDL Number:MFCD00003241