Formoxime 10% ∼2.4 mol/L in water
Supplier: TCI
Certificates
About this item
CAS Number:75-17-2
:
Formula:CH₃NO
MDL Number:MFCD00058969
:45.04 g/mol
Storage Temperature:Refrigerator
EINECS:200-845-7
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Neutralization titration):10.0 to 11.0 w/w%
- Sensitiveness:Colorimetric Test : Turn Pale Orange with Manganese
Specification Test Results
Frequently Bought Together