Flavin adenine dinucleotide disodium salt, hydrate ≥94.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:84366-81-4
:146-14-5
Formula:C₂₇H₃₃N₉O₁₅P₂
MDL Number:MFCD00041835
:829.52 g/mol
Merck Index:14,04091
Storage Temperature:Refrigerator
EINECS:282-733-8
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 94.0 area%
- Specific rotation [a]20/D:-30.0 to -25.0 deg (C=1, H2O)
- Molar extinction coefficient:min. 10,000 (H2O、448.0 to 450.0 nm)
Specification Test Results
Frequently Bought Together