9-(2-Ethylhexyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by HPLC, titration analysis)
: TCI
:476360-83-5
:
:C₃₂H₄₇B₂NO₄
:MFCD23703117
:531.35 g/mol
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Frequently Bought Together