2-[5-(2-Ethylhexyl)-2-thienyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by titrimetric analysis)
Catalog # TCIAE1295-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white clear liquid
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
About this item
:
:322.31 g/mol
Storage Temperature:Refrigerator
CAS Number:1390584-88-9
Formula:C₁₈H₃₁BO₂S