
1-Ethyl-4-methylpyridinium bromide ≥98.0% (by HPLC, titration analysis)
: TCI
:32353-49-4
:
:C₈H₁₂BrN
:MFCD18384825
:202.10 g/mol
:GHS07
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:127.0 to 131.0 deg-C
Frequently Bought Together