Order Entry
Italy
ContactUsLinkComponent
 
Ethyl-4-methyloxazole-5-carboxylate ≥96.0% (by GC)
  TCIAE1270-1G
 :  TCI
 :  
Ethyl-4-methyloxazole-5-carboxylate ≥96.0% (by GC)
  TCIAE1270-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 96.0 %

 

 

 :
 :155.15 g/mol
 :62°C
 :243-848-9
 :20485-39-6
 :C₇H₉NO₃
 :MFCD00062573