
Ethyl-2-chloropyrimidine-5-carboxylate ≥98.0% (by GC)
Catalog # TCIAE1250-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:56.0 to 60.0 deg-C
- Acid Value:max. 2.0
Specification Test Results
About this item
:
:186.60 g/mol
Boiling Pt:80°C
:GHS07
CAS Number:89793-12-4
Formula:C₇H₇ClN₂O₂
MDL Number:MFCD09863164