Ethyl-4-methylthiazole-5-carboxylate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:20582-55-2
:
Formula:C₇H₉NO₂S
MDL Number:MFCD00626873
:171.214 g/mol
Boiling Pt:233°C
Melting Pt:28°C
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together