
Ethyl-4-cyanobenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:7153-22-2
:
Formula:C₁₀H₉NO₂
MDL Number:MFCD00001824
:175.19 g/mol
Boiling Pt:140°C
EINECS:230-500-6
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:52.0 to 56.0 deg-C
Specification Test Results
Frequently Bought Together