Eprosartan Mesylate ≥98.0% (by HPLC, titration analysis)
: TCI
:144143-96-4
:
:C₂₃H₂₄N₂O₄S·CH₄O₃S
:MFCD08141807
:520.62 g/mol
:14,03635
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:246.0 to 250.0 deg-C
Frequently Bought Together