
Ethyl-2,4-dichloropyrimidine-5-carboxylate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:51940-64-8
:
Formula:C₇H₆Cl₂N₂O₂
MDL Number:MFCD09910281
:221.04 g/mol
Boiling Pt:145°C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:34.0 to 38.0 deg-C
Specification Test Results
Frequently Bought Together