
Ethyl-4-hydroxyquinoline-3-carboxylate ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:26892-90-0
:
Formula:C₁₂H₁₁NO₃
MDL Number:MFCD00173406
:217.2206 g/mol
Melting Pt:271°C
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together