Order Entry
Italy
ContactUsLinkComponent
4,4'-[1,2-Ethenediylbis(4,1-phenyleneiminocarbonyl)]bis(N-butyl-N,N-dimethylbenzenemethanaminium) Dichloride ≥94.0% (by titrimetric analysis)
Irritant
4,4'-[1,2-Ethenediylbis(4,1-phenyleneiminocarbonyl)]bis(N-butyl-N,N-dimethylbenzenemethanaminium) Dichloride ≥94.0% (by titrimetric analysis)
Catalog # TCIAE1127-1G
Supplier:  TCI
CAS Number:  
Irritant
4,4'-[1,2-Ethenediylbis(4,1-phenyleneiminocarbonyl)]bis(N-butyl-N,N-dimethylbenzenemethanaminium) Dichloride ≥94.0% (by titrimetric analysis)
Catalog # TCIAE1127-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Yellow to Green powder to crystal
  • Purity (Nonaqueous Titration):
    min. 94.0 %

Specification Test Results

About this item

 :
 :717.82 g/mol
 :GHS07
CAS Number:1296211-78-3
Formula:C₄₂H₅₄Cl₂N₄O₂