
Ethyl-1-methyl-1H-imidazole-2-carboxylate ≥98.0% (by GC, titration analysis)
: TCI
:30148-21-1
:
:C₇H₁₀N₂O₂
:MFCD02683326
:154.17 g/mol
:110°C
:GHS07
- Appearance:White to Light yellow powder to lump
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:41.0 to 48.0 deg-C
Frequently Bought Together