Order Entry
Italy
ContactUsLinkComponent
 
 
Ethyl-2-amino-4-methylthiophene-3-carboxylate ≥98.0% (by GC, titration analysis)
 :  TCI
 
 
Ethyl-2-amino-4-methylthiophene-3-carboxylate ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    79.0 to 83.0 deg-C

 

Frequently Bought Together