Order Entry
Italy
ContactUsLinkComponent
Ethyl-7-hydroxycoumarin-3-carboxylate ≥98.0% (by GC)
Ethyl-7-hydroxycoumarin-3-carboxylate ≥98.0% (by GC)
Catalog # TCIAE1017-200MG
Supplier:  TCI
CAS Number:  
Ethyl-7-hydroxycoumarin-3-carboxylate ≥98.0% (by GC)
Catalog # TCIAE1017-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    170.0 to 174.0 deg-C

Specification Test Results

About this item

 :
 :234.21 g/mol
Storage Temperature:Freezer
CAS Number:6093-71-6
Formula:C₁₂H₁₀O₅
MDL Number:MFCD00017641