
1,2-Bis(2-nitrophenoxy)ethane ≥98.0% (by GC)
: TCI
:51661-19-9
:
:C₁₄H₁₂N₂O₆
:MFCD00024226
:304.26 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:168.0 to 172.0 deg-C
Frequently Bought Together