Order Entry
Italy
ContactUsLinkComponent
Ethyl-1,3-dimethylpyrazole-5-carboxylate ≥96.0% (by GC)
Irritant
Ethyl-1,3-dimethylpyrazole-5-carboxylate ≥96.0% (by GC)
Catalog # TCIAE1001-1G
Supplier:  TCI
CAS Number:  
Irritant
Ethyl-1,3-dimethylpyrazole-5-carboxylate ≥96.0% (by GC)
Catalog # TCIAE1001-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Purity (GC):
    min. 96.0 %

Specification Test Results

About this item

 :
Boiling Pt:90°C
 :168.20 g/mol
 :GHS07
CAS Number:5744-40-1
Formula:C₈H₁₂N₂O₂
MDL Number:MFCD00233452