Ethyl pyrazole-3-carboxylate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5932-27-4
:
Formula:C₆H₈N₂O₂
MDL Number:MFCD00159643
:140.14 g/mol
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:159.0 to 163.0 deg-C
Specification Test Results
Frequently Bought Together