3-Ethyl-4-methyl-3-pyrrolin-2-one ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:766-36-9
:
Formula:C₇H₁₁NO
MDL Number:MFCD00173861
:125.17 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:102.0 to 106.0 deg-C
Specification Test Results
Frequently Bought Together