

1,2-Epoxy-4-vinylcyclohexane (mixture of isomers) ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:106-86-5
:
Formula:C₈H₁₂O
MDL Number:MFCD00022356
:124.18 g/mol
Boiling Pt:159°C
Melting Pt:–100°C
Flash Pt:58°C
Storage Temperature:Refrigerata
EINECS:203-436-1
UN:3271
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomers)
Specification Test Results
Frequently Bought Together