Inserisci ordine
Italy
ContactUsLinkComponent
 
Ethyl-2-aminothiazole-5-carboxylate ≥97.0% (by HPLC, titration analysis)
 :  TCI
 
Ethyl-2-aminothiazole-5-carboxylate ≥97.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Melting point:
    159.0 to 163.0 deg-C

 

Frequently Bought Together