
1-Ethyl-3-methylimidazolium acetate ≥95.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:143314-17-4
:
Formula:C₈H₁₄N₂O₂
MDL Number:MFCD06798186
:170.21 g/mol
Flash Pt:164°C
:GHS07
Specification Test Results
- Appearance:Light yellow to Amber to Dark green clear liquid to cloudy liquid
- Purity (HPLC):min. 95.0 % (total of Ion-Pair)
- Purity (Nonaqueous Titration):min. 95.0 %
- Specific gravity (20/20):1.0960 to 1.1060
- Refractive index n20/D:1.4940 to 1.5040
Specification Test Results
Frequently Bought Together