Ethyl (R)-4-phenyl-2-(trifluoromethanesulphonyloxy)butyrate ≥97.0% (by GC)
Catalog # TCIAE0879-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
About this item
:
:340.31 g/mol
CAS Number:88767-98-0
Formula:C₁₃H₁₅F₃O₅S
MDL Number:MFCD09751096