



Ethylmagnesium chloride ∼1.0 mol/L in THF activated with Zinc Chloride (∼10mol%), activated with zinc chloride (ca. 10mol%)
Catalog # TCIAE0778-250G
Supplier: TCI
CAS Number:
Specifications
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Concentration (Neutralization titration):9.0 to 11.0 w/w%
Specification Test Results
About this item
:
:88.82 g/mol
EINECS:219-206-9
UN:3399
ADR:4.3,II
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:2386-64-3
Formula:C₂H₅ClMg
MDL Number:MFCD00000472