
1-Ethyl-3-methylpyridinium bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, titration analysis)
TCIAE0756-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 98.0 %
:
:402.33 g/mol
:GHS07
:Refrigerator
:841251-37-4
:C₁₀H₁₂F₆N₂O₄S₂