Order Entry
Italy
ContactUsLinkComponent
 
 
7-Ethyl-10-hydroxycamptothecin ≥98.0% (by HPLC)
 :  TCI
 
undefined
 
7-Ethyl-10-hydroxycamptothecin ≥98.0% (by HPLC)
 :  TCI

 

  • Appearance:
    White to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 94.0 %
  • Specific rotation [a]20/D:
    +20.0 to +30.0 deg (C=0.2, THF)

 

Frequently Bought Together