Order Entry
Italy
ContactUsLinkComponent
Ethyl-2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate ≥98.0% (by titrimetric analysis)
Ethyl-2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate ≥98.0% (by titrimetric analysis)
Catalog # TCIAE0667-1G
Supplier:  TCI
CAS Number:  
Ethyl-2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate ≥98.0% (by titrimetric analysis)
Catalog # TCIAE0667-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :262.11 g/mol
Storage Temperature:Refrigerator
CAS Number:346656-34-6
Formula:C₁₄H₁₉BO₄
MDL Number:MFCD08276372