
EDTA tetrasodium salt tetrahydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13235-36-4
:
Formula:C₁₀H₁₂N₂O₈Na₄·4H₂O
MDL Number:MFCD00150027
:452.23 g/mol
:GHS07
Specification Test Results
- Appearance:White powder to crystal
- Purity (Chelometric Titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:14.0 to 18.0 %
Specification Test Results
Frequently Bought Together