Order Entry
Italy
ContactUsLinkComponent
Ethyl-3-ethoxy-2,2-difluoro-3-hydroxypropionate ≥83.0% (by GC)
Ethyl-3-ethoxy-2,2-difluoro-3-hydroxypropionate ≥83.0% (by GC)
  TCIAE0547-1G
 :  TCI
 :  
Ethyl-3-ethoxy-2,2-difluoro-3-hydroxypropionate ≥83.0% (by GC)
  TCIAE0547-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 83.0 %

 

 

 :
 :198.17 g/mol
 :Refrigerator
 :141546-97-6
 :C₇H₁₂F₂O₄
 :MFCD00718642