3,4-Ethylenedioxynitrobenzene
5 g
Codice catalogo TCIAE0523-5G
Nome del fornitore: TCI
3,4-Ethylenedioxynitrobenzene
Codice catalogo TCIAE0523-5G
Nome del fornitore: TCI
Numero CAS: 16498-20-7
Jump to:
Product Details & Documents
Synonyms: 6-Nitro-2,3-dihydrobenzo[b][1,4]dioxine, 6-Nitro-1,4-benzodioxane
- CAS Number:16498-20-7
- Formula:C₈H₇NO₄
- MDL Number:MFCD00088689
- Molecular Weight:181.15 g/mol
- EINECS:240-565-2
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:119.0 to 122.0 deg-C
- Solubility in hot Methanol:almost transparency
Specifications
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



