
4,4'-Ethylidenebisphenol ≥98.0% (by GC)
Catalog # TCIAE0432-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:125.0 to 128.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:214.26 g/mol
:GHS07
CAS Number:2081-08-5
:MFCD00186869
Formula:C₁₄H₁₄O₂
MDL Number:MFCD00191431