Order Entry
Italy
ContactUsLinkComponent
Ethylene glycolbis(4-carboxyphenyl)ether ≥95.0% (by titrimetric analysis)
Ethylene glycolbis(4-carboxyphenyl)ether ≥95.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Ethylene glycolbis(4-carboxyphenyl)ether ≥95.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 95.0 %

Specification Test Results

Related Information

Frequently Bought Together