
Eicosanedioic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2424-92-2
:
Formula:C₂₀H₃₈O₄
MDL Number:MFCD00059627
Boiling Pt:234°C
:342.52 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 94.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:124.0 to 128.0 deg-C
Specification Test Results
Frequently Bought Together