
Potassium-2-ethylhexanoate ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3164-85-0
:
Formula:C₈H₁₆O₂
MDL Number:MFCD00045896
:182.30 g/mol
EINECS:221-625-7
:GHS07
Specification Test Results
- Appearance:White or Colorless powder to lump
- Purity (Nonaqueous Titration):min. 95.0 %
- Water:max. 5.0 %
Specification Test Results
Frequently Bought Together