
2,6-DBEAQ ≥95.0% (by HPLC, titration analysis)
Catalog # TCIAD5764-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystaline
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 95.0 %
Specification Test Results
About this item
:
:412.39 g/mol
:GHS07
CAS Number:2370885-20-2
Formula:C₂₂H₂₀O₈