


Difluoroiodomethane ∼2 mol/L in acetonitrile
TCIAD5762-5ML
: TCI
:
- Pk:5 mL
- Seq:2210
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (HNMR):1.8 to 2.2 mol/L
:
:1648
:3,II
:GHS02
:GHS06
:GHS08
:Freezer
:1493-03-4
:CHF₂I
:MFCD03094113