
N,2-Diphenyl[60]fulleropyrrolidine ≥98.0% (by HPLC) (contains 5% Hexane at maximum)
Catalog # TCIAD5757-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Orange to Amber to Dark red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Hexane:max. 5.0 %
Specification Test Results
About this item
:
:915.93 g/mol
:GHS08
CAS Number:1373934-14-5
Formula:C₇₄H₁₃N