Order Entry
Italy
ContactUsLinkComponent
 
10-[4-[4,6-Di(adamantan-1-yl)-1,3,5-triazin-2-yl]phenyl]-9,9-diphenyl-9,10-dihydroacridine ≥99.0% (by HPLC), purified by sublimation
Catalog # TCIAD5746-200MG
Supplier:  TCI
CAS Number:  
10-[4-[4,6-Di(adamantan-1-yl)-1,3,5-triazin-2-yl]phenyl]-9,9-diphenyl-9,10-dihydroacridine ≥99.0% (by HPLC), purified by sublimation
Catalog # TCIAD5746-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystaline
  • Purity (HPLC):
    min. 99.0 area%

Specification Test Results

About this item

 :
 :757.04 g/mol
CAS Number:2250187-17-6
Formula:C₅₄H₅₂N₄